siRNA Details
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027022.4 | NP_081298.1 | 4 | 579 | ||
GenBank mRNA | AK005603.1 | - | 4 | 532 | ||
AK161303.1 | - | 4 | 573 | |||
AY172740.1 | - | 4 | 460 | |||
AY241871.1 | - | 4 | 579 | |||
BC049747.1 | - | 4 | 572 | 6772661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
