siRNA Details
Gene Symbol: Fam177a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028527.1 | NP_082803.1 | 1, 2 | 200 | ||
GenBank mRNA | AK006713.1 | - | 33 | |||
AK045530.1 | - | 260 | ||||
AK159923.1 | - | 238 | ||||
BC048158.1 | - | 200 | 5698044 | |||
BC049669.1 | - | 259 | 6535412 |
Gene Symbol: 1700047I17Rik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100116.1 | NP_001093586.1 | 200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
