siRNA Details
Gene Symbol: Bpifa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291079.1 | NP_001278008.1 | 3, 4 | 596 | ||
NM_028528.3 | NP_082804.1 | 3, 4 | 596 | |||
GenBank mRNA | AK006829.1 | - | 3, 4 | 586 | ||
BC049691.1 | - | 3, 4 | 585 | 6705503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
