siRNA Details
Gene Symbol: 1700065D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006889.1 | - | 1 | 403 | ||
AK045280.1 | - | 1 | 645 | |||
BC053441.1 | - | 1 | 852 | 6332684 | ||
BF018438.1 | - | 1 | 49 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
