siRNA Details
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028536.1 | NP_082812.1 | 3 | 884 | ||
GenBank mRNA | AK006786.1 | - | 2 | 276 | ||
AK086180.1 | - | 2 | 589 | |||
BC024705.1 | - | 2 | 541 | 3668303 | ||
BC036300.1 | - | 3 | 884 | 4501054 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
