siRNA Details
Gene Symbol: Tex35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006825.1 | - | 5 | 304 | ||
BC099580.1 | - | 5 | 299 | 30920417 | ||
BC115790.1 | - | 5 | 301 | 40100579 | ||
BY080014.1 | - | 4 | 199 | |||
BY706810.1 | - | 5 | 304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
