siRNA Details
Gene Symbol: Als2cr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175200.4 | NP_780409.2 | 11, 12 | 1310 | ||
GenBank mRNA | AK076617.1 | - | 11, 12 | 1312 | ||
BC049729.1 | - | 7, 8 | 527 | |||
BC118929.2 | - | 11, 12 | 1276 | 40090362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
