siRNA Details
Gene Symbol: 1700066M21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028546.1 | NP_082822.1 | 2 | 380 | ||
GenBank mRNA | CJ047538.1 | - | 2 | 382 |

More Information
Other Species Targeted

Gene Ontology Categories:
