siRNA Details
Gene Symbol: Camkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028576.2 | NP_082852.1 | 2 | 309 | ||
GenBank mRNA | AK007130.1 | - | 2 | 404 | ||
AK155246.1 | - | 2 | 310 | |||
AK172547.1 | - | 2 | 255 | |||
BC138272.1 | - | 2 | 457 | 8861291 | ||
BC138273.1 | - | 2 | 457 | 8861292 |
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147783.1 | - | 346 |
