siRNA Details
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA427326.1 | - | 5 | 475 | ||
AK008857.1 | - | 4 | 327 | |||
AW105955.1 | - | 4 | 358 | |||
BC021953.1 | - | 6 | 683 | |||
BC099512.1 | - | 5 | 442 | |||
BY261359.1 | - | 4 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
