siRNA Details
Gene Symbol: Cabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144532.2 | NP_653115.1 | 1 | 96 | ||
GenBank mRNA | AK171863.1 | - | 5 | 706 | ||
AY039218.1 | - | 1 | 91 | |||
BC049263.1 | - | 1 | 96 | 5366999 | ||
BC053330.1 | - | 1 | 51 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
