siRNA Details
Gene Symbol: Defb30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039566.2 | NP_001034655.1 | 2 | 164 | ||
GenBank mRNA | AK010780.1 | - | 271 | |||
AK019622.1 | - | 275 | ||||
AK078987.1 | - | 2 | 153 | |||
BC132184.1 | - | 2 | 151 | 40130461 | ||
BC132214.1 | - | 2 | 151 | 40130491 | ||
DQ012040.1 | - | 2 | 151 | |||
DQ141309.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
