siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 2, 3 | 226 | ||
NM_028610.2 | NP_082886.2 | 2, 3 | 226 | |||
GenBank mRNA | AF490348.1 | - | 2, 3 | 143 | ||
AK010753.1 | - | 2, 3 | 194 | |||
AK033197.1 | - | 2, 3 | 227 | |||
BC064754.1 | - | 2, 3 | 165 | 30024281 | ||
BY731814.1 | - | 2, 3 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
