siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 4 | 576 | ||
NM_028610.2 | NP_082886.2 | 5 | 732 | |||
GenBank mRNA | AF490348.1 | - | 5 | 649 | ||
AK033197.1 | - | 5 | 733 | |||
BC064754.1 | - | 5 | 671 | 30024281 | ||
BY731814.1 | - | 4 | 542 |
