siRNA Details
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018002.1 | NP_001018002.1 | 4 | 565 | ||
NM_028610.2 | NP_082886.2 | 5 | 721 | |||
GenBank mRNA | AF490348.1 | - | 5 | 638 | ||
AK033197.1 | - | 5 | 722 | |||
BC064754.1 | - | 5 | 660 | 30024281 | ||
BY731814.1 | - | 4 | 531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
