siRNA Details
Gene Symbol: Ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028611.3 | NP_082887.2 | 10 | 1172 | ||
GenBank mRNA | AK010752.1 | - | 10 | 1171 | ||
AK090097.1 | - | 9 | 1578 | |||
BC028524.1 | - | 3 | 411 | |||
BC108350.1 | - | 9 | 1566 | 4220568 | ||
BC116213.1 | - | 3 | 264 | |||
BC116214.1 | - | 3 | 264 | |||
BC128473.1 | - | 3 | 264 | |||
BC128474.1 | - | 3 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
