siRNA Details
Gene Symbol: Ppp2r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034085.2 | NP_001029257.1 | 13 | 1748 | ||
NM_001286553.1 | NP_001273482.1 | 13 | 1748 | |||
NM_028614.3 | NP_082890.2 | 13 | 1748 | |||
GenBank mRNA | AK010754.1 | - | 4 | 405 | ||
AK077158.1 | - | 13 | 1702 | |||
AK161392.1 | - | 13 | 1701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
