siRNA Details
Gene Symbol: Tubb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023716.2 | NP_076205.1 | 2, 3 | 285 | ||
GenBank mRNA | AK010786.1 | - | 2, 3 | 286 | ||
AK168908.1 | - | 2, 3 | 291 | |||
BC138935.1 | - | 2, 3 | 272 | 8861957 | ||
BC138936.1 | - | 2, 3 | 272 | 8861958 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
