siRNA Details
Gene Symbol: Cby1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028634.3 | NP_082910.1 | 4 | 379 | ||
GenBank mRNA | AF331040.1 | - | 4 | 379 | ||
AK003719.1 | - | 4 | 355 | |||
BC005733.1 | - | 3 | 332 | 3596486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
