siRNA Details
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008791.2 | NP_001008791.1 | 12 | 3244 | ||
NM_001008792.2 | NP_001008792.1 | 11 | 3223 | |||
NM_001008793.2 | NP_001008793.1 | 11 | 3208 | |||
NM_028640.2 | NP_082916.1 | 11 | 3211 | |||
GenBank mRNA | AK122523.1 | - | 11 | 2339 | ||
AY739114.1 | - | 11 | 3171 | |||
AY739115.1 | - | 12 | 3204 | |||
AY739116.1 | - | 11 | 3183 | |||
AY739117.1 | - | 11 | 3168 | |||
AY739118.1 | - | 8 | 1820 | |||
AY739119.1 | - | 7 | 1787 | |||
AY739120.1 | - | 6 | 1574 | |||
AY739121.1 | - | 6 | 1692 | |||
AY739122.1 | - | 7 | 2024 | |||
BC150757.1 | - | 11 | 2568 | 9087668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
