siRNA Details
Gene Symbol: Prr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163551.1 | NP_001157023.1 | 5 | 324 | ||
NM_028648.3 | NP_082924.2 | 4 | 300 | |||
GenBank mRNA | AK015291.1 | - | 4 | 301 | ||
BC049645.1 | - | 5 | 334 | |||
BC103787.1 | - | 5 | 335 | 30919261 | ||
BY714849.1 | - | 4 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
