siRNA Details
Gene Symbol: Fam83e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033170.4 | NP_001028342.2 | 2 | 1532 | ||
GenBank mRNA | AK144522.1 | - | 2 | 1534 | ||
BC042793.1 | - | 2 | 379 | |||
BC139284.1 | - | 2 | 1452 | 8862306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
