siRNA Details
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285942.1 | NP_001272871.1 | 4 | 467 | ||
NM_001285943.1 | NP_001272872.1 | 4 | 467 | |||
NM_028659.3 | NP_082935.1 | 5 | 566 | |||
GenBank mRNA | AK004664.1 | - | 5 | 493 | ||
AK037834.1 | - | 5 | 462 | |||
AK158875.1 | - | 4 | 444 | |||
BC027638.1 | - | 4 | 366 | 1348838 | ||
BC091749.1 | - | 5 | 470 | 3661337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
