siRNA Details
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028659.3 | NP_082935.1 | 2 | 251 | ||
GenBank mRNA | AK004664.1 | - | 2 | 178 | ||
AK037834.1 | - | 2 | 147 | |||
BC091749.1 | - | 2 | 155 | 3661337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
