siRNA Details
Gene Symbol: Atp6v1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023721.2 | NP_076210.1 | 5 | 515 | ||
GenBank mRNA | AF298810.1 | - | 5 | 336 | ||
AK003446.1 | - | 2 | 52 | |||
AK146129.1 | - | 3 | 186 | |||
AK146448.1 | - | 5 | 418 | |||
AK147910.1 | - | 432 | ||||
AK150981.1 | - | 5 | 404 | |||
AK152367.1 | - | 5 | 404 | |||
AK153250.1 | - | 5 | 404 | |||
AK165761.1 | - | 5 | 409 | |||
BC033457.1 | - | 5 | 377 | 3991107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
