siRNA Details
Gene Symbol: Ankrd42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028665.4 | NP_082941.2 | 7 | 1830 | ||
GenBank mRNA | AK016847.1 | - | 2 | 111 | ||
AK029511.1 | - | 7 | 1832 | |||
AK029888.1 | - | 7 | 1791 | |||
AK033599.1 | - | 7 | 1819 | |||
AK048925.1 | - | 7 | 1724 | |||
AK133339.1 | - | 1791 | ||||
BC030885.1 | - | 7 | 1459 | |||
BC100433.1 | - | 7 | 1866 | |||
BC145456.1 | - | 7 | 948 | 9054017 | ||
BC145457.1 | - | 6 | 812 | |||
BC145458.1 | - | 6 | 812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
