siRNA Details
Gene Symbol: D3Ertd751e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099785.1 | NP_001093255.1 | 5 | 378 | ||
NM_028667.3 | NP_082943.2 | 6 | 495 | |||
GenBank mRNA | AK015146.1 | - | 5 | 449 | ||
AK039725.1 | - | 5 | 342 | |||
AK045813.1 | - | 5 | 342 | |||
AK085404.1 | - | 5 | 376 | |||
AK138405.1 | - | 7 | 600 | |||
AK144064.1 | - | 5 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
