siRNA Details
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285959.1 | NP_001272888.1 | 2 | 140 | ||
NM_001285960.1 | NP_001272889.1 | 2 | 273 | |||
NM_029610.3 | NP_083886.1 | 2 | 524 | |||
GenBank mRNA | AK009135.1 | - | 2 | 527 | ||
AK015083.1 | - | 3 | 332 | |||
AK157931.1 | - | 142 | ||||
BC011339.1 | - | 2 | 314 | 3978191 | ||
BY752674.1 | - | 2 | 142 | |||
DV043140.1 | - | 1 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
