siRNA Details
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081164.1 | NP_001074633.1 | 7 | 786 | ||
NM_001256033.1 | NP_001242962.1 | 7 | 786 | |||
GenBank mRNA | AK122429.1 | - | 2 | 77 | ||
BC138373.1 | - | 7 | 758 | 8861393 | ||
BC145259.1 | - | 7 | 758 | 9053812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
