siRNA Details
Gene Symbol: Als2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159948.2 | NP_001153420.2 | 22 | 3877 | ||
NM_028717.6 | NP_082993.4 | 22 | 3708 | |||
GenBank mRNA | AB053307.1 | - | 22 | 3696 | ||
AK014320.1 | - | 10 | 1037 | |||
AK029858.1 | - | 22 | 3881 | |||
AK171800.1 | - | 7 | 738 | |||
BC046828.1 | - | 22 | 3669 | 6409924 | ||
CN715620.1 | - | 1 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
