siRNA Details
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028719.1 | NP_082995.1 | 5 | 762 | ||
GenBank mRNA | AK014396.1 | - | 5 | 764 | ||
AK043692.1 | - | 5 | 775 | |||
BC043087.1 | - | 5 | 762 | 6417611 | ||
BC063081.1 | - | 5 | 762 | 6417611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
