siRNA Details
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028719.1 | NP_082995.1 | 5, 6 | 815 | ||
GenBank mRNA | AK014396.1 | - | 5, 6 | 817 | ||
AK043692.1 | - | 5, 6 | 828 | |||
AK202965.1 | - | 3, 4 | 181 | |||
BC043087.1 | - | 5, 6 | 815 | 6417611 | ||
BC063081.1 | - | 5, 6 | 815 | 6417611 |

More Information
Other Species Targeted

Gene Ontology Categories:
