siRNA Details
Gene Symbol: 4632428N05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159572.1 | NP_001153044.1 | 2 | 613 | ||
NM_028732.4 | NP_083008.1 | 2 | 613 | |||
GenBank mRNA | AK014600.1 | - | 2 | 541 | ||
AK030479.1 | - | 2 | 553 | |||
AK032383.1 | - | 2 | 558 | |||
AK033393.1 | - | 2 | 559 | |||
AK079734.1 | - | 2 | 561 | |||
AK131118.1 | - | 2 | 529 | |||
AK159086.1 | - | 2 | 558 | |||
AK159864.1 | - | 2 | 558 | |||
AK162635.1 | - | 452 | ||||
AK163118.1 | - | 2 | 558 | |||
AK170059.1 | - | 2 | 561 | |||
AK170419.1 | - | 2 | 558 | |||
BC003967.1 | - | 2 | 529 | 3488482 | ||
EF426799.1 | - | 2 | 442 | |||
JN602184.1 | - | 2 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
