siRNA Details
Gene Symbol: Serpina3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167705.1 | NP_001161177.1 | 5 | 1388 | ||
NM_028740.2 | NP_083016.1 | 4 | 728 | |||
GenBank mRNA | AK016702.1 | - | 4 | 731 | ||
BC063325.1 | - | 5 | 1578 | 30280343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
