siRNA Details
Gene Symbol: Ifltd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028742.2 | NP_083018.1 | 9 | 1333 | ||
GenBank mRNA | AK016641.1 | - | 9 | 1333 | ||
AK035947.1 | - | 1070 | ||||
AK052206.1 | - | 7 | 1412 | |||
AY753326.1 | - | 7 | 1168 | |||
BC053420.1 | - | 8 | 1295 | 6705208 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
