siRNA Details
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028758.2 | NP_083034.1 | 17 | 1829 | ||
GenBank mRNA | AK004632.1 | - | 17 | 1801 | ||
AK034612.1 | - | 14 | 1472 | |||
AK129280.1 | - | 17 | 1788 | |||
AK143691.1 | - | 17 | 1817 | |||
AK149798.1 | - | 17 | 1819 | |||
AK154904.1 | - | 17 | 1804 | |||
AK159953.1 | - | 17 | 1819 | |||
AK180517.1 | - | 2 | 107 | |||
BC057377.1 | - | 10 | 1332 | 5698559 | ||
BC062933.1 | - | 17 | 1798 | 30356060 |
