siRNA Details
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028766.2 | NP_083042.1 | 10, 11 | 1026 | ||
GenBank mRNA | AK004778.1 | - | 10, 11 | 1027 | ||
AK037165.1 | - | 10, 11 | 997 | |||
AK050173.1 | - | 10, 11 | 1066 | |||
AK151693.1 | - | 10, 11 | 1019 | |||
AK151766.1 | - | 10, 11 | 1044 | |||
AK152078.1 | - | 10, 11 | 1019 | |||
AK168559.1 | - | 10, 11 | 1034 | |||
BC005521.1 | - | 10, 11 | 992 | |||
BC024933.1 | - | 10, 11 | 1013 | 3990353 |
