siRNA Details
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028766.2 | NP_083042.1 | 10, 11 | 1021 | ||
GenBank mRNA | AK004778.1 | - | 10, 11 | 1022 | ||
AK037165.1 | - | 10, 11 | 992 | |||
AK050173.1 | - | 10, 11 | 1061 | |||
AK151693.1 | - | 10, 11 | 1014 | |||
AK151766.1 | - | 10, 11 | 1039 | |||
AK152078.1 | - | 10, 11 | 1014 | |||
AK168559.1 | - | 10, 11 | 1029 | |||
BC005521.1 | - | 10, 11 | 987 | |||
BC024933.1 | - | 10, 11 | 1008 | 3990353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
