siRNA Details
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028773.3 | NP_083049.1 | 2 | 270 | ||
GenBank mRNA | AJ306422.1 | - | 2 | 136 | ||
AK004734.2 | - | 2 | 271 | |||
AK156202.1 | - | 2 | 335 | |||
AK171017.1 | - | 2 | 268 | |||
BC064734.1 | - | 2 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
