siRNA Details
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166506.1 | NP_001159978.1 | 12 | 1564 | ||
NM_001166507.1 | NP_001159979.1 | 12 | 1564 | |||
NM_028777.3 | NP_083053.2 | 12 | 1564 | |||
GenBank mRNA | AK004818.1 | - | 12 | 1489 | ||
AK047396.1 | - | 12 | 1572 | |||
AK172947.1 | - | 9 | 1099 | |||
AK193303.1 | - | 3 | 243 | |||
BC005766.1 | - | 12 | 1522 | 2649054 | ||
BC086620.1 | - | 12 | 1498 |
