siRNA Details
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 24 | 3851 | ||
GenBank mRNA | AB093256.1 | - | 21 | 3510 | ||
AK004841.1 | - | 12 | 1399 | |||
AK036540.1 | - | 24 | 3835 | |||
AK037082.1 | - | 9 | 931 | |||
AK145708.1 | - | 23 | 3740 | |||
AK150045.1 | - | 24 | 3811 | |||
AK154925.1 | - | 24 | 3852 | |||
AK157310.1 | - | 24 | 3861 | |||
AK180241.1 | - | 1 | 90 | |||
AK184296.1 | - | 1 | 70 | |||
AK194929.1 | - | 1 | 28 | |||
AK201971.1 | - | 2 | 145 | |||
BC043323.1 | - | 6 | 692 | |||
BC060069.1 | - | 19 | 2963 | |||
BC061500.1 | - | 2 | 181 | |||
BC072573.1 | - | 22 | 4550 | 5705116 | ||
BC094446.1 | - | 15 | 2431 |

More Information
Other Species Targeted

Gene Ontology Categories:
