siRNA Details
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023739.3 | NP_076228.2 | 20 | 2982 | ||
GenBank mRNA | AF223576.1 | - | 20 | 2946 | ||
AK007054.1 | - | 2 | 102 | |||
AK155387.1 | - | 20 | 2934 |
