siRNA Details
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028797.1 | NP_083073.1 | 7 | 519 | ||
GenBank mRNA | AK005675.1 | - | 7 | 497 | ||
BU945975.1 | - | 7 | 521 | |||
CO812705.1 | - | 7 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
