siRNA Details
Gene Symbol: Crct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028798.3 | NP_083074.2 | 2 | 532 | ||
GenBank mRNA | AK009010.1 | - | 2 | 533 | ||
BC060280.1 | - | 2 | 511 | 30290660 | ||
CB202103.1 | - | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
