siRNA Details
Gene Symbol: Rnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028810.2 | NP_083086.1 | 1 | 294 | ||
GenBank mRNA | AK011442.1 | - | 1 | 292 | ||
AK035195.1 | - | 1 | 279 | |||
AK048635.1 | - | 1 | 292 | |||
BC009002.1 | - | 1 | 264 | 3500586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
