siRNA Details
Gene Symbol: Ttc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152817.4 | NP_690030.3 | 16 | 2045 | ||
GenBank mRNA | AK012131.1 | - | 16 | 2045 | ||
AK030942.1 | - | 16 | 2046 | |||
AK049324.1 | - | 16 | 2046 | |||
AK168138.1 | - | 16 | 2036 | |||
BC021912.1 | - | 15 | 1780 | 4503335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
