siRNA Details
Gene Symbol: 2810403A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028814.3 | NP_083090.2 | 10 | 1124 | ||
GenBank mRNA | AK012969.1 | - | 10 | 1101 | ||
AK083018.1 | - | 10 | 1085 | |||
AK170490.1 | - | 10 | 1119 | |||
AK173057.1 | - | 10 | 1088 | |||
BC006621.1 | - | 10 | 1099 | 3158104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
