siRNA Details
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033606.2 | NP_001028778.2 | 13, 14 | 2172 | ||
NM_001136222.1 | NP_001129694.1 | 12, 13 | 1754 | |||
NM_028817.3 | NP_083093.2 | 14, 15 | 2312 | |||
GenBank mRNA | AK012088.1 | - | 13, 14 | 2115 | ||
AK160839.1 | - | 12, 13 | 1698 | |||
AK161708.1 | - | 14, 15 | 2313 | |||
AK166723.1 | - | 14, 15 | 2250 | |||
BC031529.1 | - | 12, 13 | 1897 | 4009004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
