siRNA Details
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033606.2 | NP_001028778.2 | 15 | 2416 | ||
NM_001136222.1 | NP_001129694.1 | 14 | 1998 | |||
NM_028817.3 | NP_083093.2 | 16 | 2556 | |||
GenBank mRNA | AK012088.1 | - | 15 | 2359 | ||
AK160839.1 | - | 14 | 1942 | |||
AK161708.1 | - | 16 | 2557 | |||
AK166723.1 | - | 16 | 2496 | |||
BC031529.1 | - | 14 | 2141 | 4009004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
