siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 2 | 2169 | ||
GenBank mRNA | AK029230.1 | - | 2173 | |||
AK031730.1 | - | 2 | 2172 | |||
AK044396.1 | - | 2 | 2176 | |||
AK080676.1 | - | 2 | 2182 | |||
AK155015.1 | - | 2 | 2146 | |||
AK165963.1 | - | 2 | 2170 | |||
BC118527.1 | - | 2 | 2023 | 40098810 |
